Learn More
Thermo Scientific™ Barnstead™ Classic Stills Replacement Consumables
GSA_VA
Catalog No. 0902845
Change view
Click to view available options
Type:
Replacement Filter
Replacement UV Lamp
For Use With (Equipment):
Barnstead Classic Stills
Barnstead Classic Stills Reservoirs, H4005
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type For Use With (Equipment)
09-028-45 Replacement UV Lamp Barnstead Classic Stills Reservoirs, H4005
09-050-116 Replacement Filter Barnstead Classic Stills
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09-028-45 Supplier Thermo Scientific™ Supplier No. 04141
Add to Cart
Add to Cart
Maintain the performance of your Thermo Scientific™ Barnstead™ Classic Still with replacement consumables such as UV lamps and air filters.
Maintain the performance of your still with Thermo Scientific™ Barnstead™ Classic Stills Replacement Consumables, including UV lamp and Thermo Scientific™ Ventgard™ filter element (the consumable portion of the Ventgard filter assembly). Prevent air from entering the reservoir via the condenser atmospheric vent and protect stored water against airborne particulates, organics and CO2 with the Ventgard air filter.
Maintain the performance of your still with Thermo Scientific™ Barnstead™ Classic Stills Replacement Consumables, including UV lamp and Thermo Scientific™ Ventgard™ filter element (the consumable portion of the Ventgard filter assembly). Prevent air from entering the reservoir via the condenser atmospheric vent and protect stored water against airborne particulates, organics and CO2 with the Ventgard air filter. 
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Type Replacement UV Lamp
For Use With (Equipment) Barnstead Classic Stills Reservoirs, H4005