Description
Replace worn, outdated and otherwise ineffective cartridges with Thermo Scientific™ Barnstead™ E-Pure™ Cartridge Kits. These cartridge kits are classified based on type of feed water used; choose the appropriate kit according to your feed water and need for organic free water.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Description | Cartridge kit for E-Pure 4-holder; Pretreated Feed |
| Feedwater Source | Pretreated |
| For Use With (Equipment) | E-Pure 4-holder |
| Type | Cartridge Kit |