Learn More
Thermo Scientific™ Barnstead™ E-Pure™ Cartridge Kits
GSA_VA
Catalog No. p-4531451
Change view
Click to view available options
For Use With (Equipment):
E-Pure 3-holder
E-Pure 3-holder, Organic free
E-Pure 4-holder
E-Pure 4-holder, Organic free
E-Pure Ultrapure Water Purification System
Feedwater Source:
N/A
Pretreated
Pretreated feed
Tap
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Feedwater Source
09-050-268 E-Pure 3-holder, Organic free Pretreated feed
19-025-470 E-Pure 4-holder Pretreated
09-050-285 E-Pure 4-holder, Organic free Pretreated
09-050-269 E-Pure 4-holder, Organic free Tap
09-050-277 E-Pure 4-holder Tap
09-050-278 E-Pure 3-holder Pretreated feed
09-034-15 E-Pure Ultrapure Water Purification System N/A
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 09-050-268 Supplier Thermo Scientific™ Supplier No. D5022
Only null left
Add to Cart
Add to Cart
Replace worn, outdated and otherwise ineffective cartridges with Thermo Scientific™ Barnstead™ E-Pure™ Cartridge Kits.
Replace worn, outdated and otherwise ineffective cartridges with Thermo Scientific™ Barnstead™ E-Pure™ Cartridge Kits. These cartridge kits are classified based on type of feed water used; choose the appropriate kit according to your feed water and need for organic free water.
Replace worn, outdated and otherwise ineffective cartridges with Thermo Scientific™ Barnstead™ E-Pure™ Cartridge Kits. These cartridge kits are classified based on type of feed water used; choose the appropriate kit according to your feed water and need for organic free water.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Description Cartridge kit for E-Pure 3-holder, Pretreated feed, Organic
Feedwater Source Pretreated feed
For Use With (Equipment) E-Pure 3-holder, Organic free
Type Cartridge Kit