Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Barnstead™ E-Pure™ Ultrapure Water Purification Systems
GSA_VA

Catalog No. 09050254 Shop All Thermo Scientific Products
Change view
Click to view available options
Modules:
3
4
Voltage:
120 V
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Modules Voltage
09-050-254 3 120 V
09-050-255 4 120 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09-050-254 Supplier Thermo Scientific™ Supplier No. D4631
  • $9,340.00
    $7,352.65 / Each of 1

    Save $1,987.35

    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Produce Type 1 ultrapure water with low operating costs using the Thermo Scientific™ Barnstead™ E-Pure™ Ultrapure Water Purification system.

Produce Type 1 ultrapure water with low operating costs using Thermo Scientific™ Barnstead™ E-Pure™ Ultrapure Water Purification Systems. Choose from a three- or four-module system. Gain longer cartridge life and benefit from lower operating costs with large cartridges. This system is Ideal for AA, GS, buffer preps, or as a purified feed water source for analytical instruments or labware washers. It is simple to use and easy to maintain.

AquaPro™ digital on-line analytical measurement platform featuring:

  • Large color graphic display
  • Temperature readout
  • Temperature compensated
  • Calibratable

Features:
  • System achieves resistivities up to 18.2 MΩ-cm and TOC levels below 10ppb.
  • Virgin polypropylene water pathways prevent recontamination.
  • Recirculation pump continuously recirculates water throughout the entire system to maintain water purity and reduce the quantity of wastewater required for rinse-up.
  • System is wall-mountable and completely assembled for easy installation.
  • Quarter-turn quick release canisters allow for easy cartridge replacement.
  • 0.2μm absolute final filter removes bacteria and particulates that could alter your results.
  • Digital resistivity meter is automatically temperature-compensated to 25°C, preventing fluctuating readings due to temperature changes.

Ordering Information:

Optional accessories include Remote Dispenser (D8952), Low Pressure Pump Protector (D2706), and Low Water Level Pump Protector (D0603, D0606). These products, as well as replacement cartridges and consumables, are available separately. For details, contact your sales representative.

Includes:

Wall bracket

Requires:

O-rings included with cartridge pack (sold separately)

Recommended for:

AA, GS, buffer preps, feedwater for analytical instruments or labware washers

Note:

If feeding E-Pure from a storage reservoir, Low Water Pump Protector is recommended to protect pump from damage if reservoir water level falls below system requirements. Order separately (D0603 for 120V, D0606 for 240V).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, CSA
Conductivity Type 1: 0.055μS/cm, Type 2: 0.067 to 0.1μS/cm
Bacterial Content <1 CFU/mL
Feed Water Monitoring No
Feedwater Source Tap or Pretreated
Flow Rate Up to 2.5 L/min.
Product Water Type Type 1
Remote Dispenser Optional
Resistivity 18.2 Mohm.cm at 25°C
TOC Monitor with UV Intensity Monitoring No
Total Organic Carbon <10 ppb
Feedwater Pressure 100 psig (6.9 bar)
Modules 3
Particles >0.2µM/mL <1 μM/mL
Dimensions (L x W x H) 7.5 x 31.5 x 28 in. (19 x 80.0 x 71.1 cm)
Length (English) 7.5 in.
Length (Metric) 19 cm
Width (English) 31.5 in.
Width (Metric) 80.0 cm
Height (English) 28 in.
Height (Metric) 71.1 cm
Voltage 120 V
Product Type Water Purification System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.