Learn More
Thermo Scientific™ Barnstead™ Easypure™ Cartridges and Filters
GSA_VA
Catalog No. 09034166
Change view
Click to view available options
For Use With (Equipment):
EasyPure II RF
Easypure II
Easypure II UV, Easypure II UV/UF
Easypure II, Easypure UF
Easypure UF, Easypure II UV/UF
Type:
Cartridge Pack, Deionized Feed
Cartridge Pack, RO or Distilled Feed
Cartridge and Filter
Sanitation Cartridge
Ultrafilter
Resin Type:
2/3 Macroreticular Resin
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Type Resin Type
09-034-166 Easypure II UV, Easypure II UV/UF Cartridge Pack, RO or Distilled Feed -
09-034-165 Easypure II UV, Easypure II UV/UF Cartridge Pack, Deionized Feed -
09-034-167 Easypure II, Easypure UF Cartridge Pack, Deionized Feed -
13-687-217 Easypure UF, Easypure II UV/UF Ultrafilter -
09-034-17 Easypure II Cartridge and Filter 2/3 Macroreticular Resin
09-050-66 EasyPure II RF Sanitation Cartridge -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 09-034-166 Supplier Thermo Scientific™ Supplier No. D502125
Extend the life of your Thermo Scientific™ Barnstead™ Easypure™ II or Easypure™ RF water system with replacement cartridges and filters.
Extend the life of your Thermo Scientific™ Barnstead™ Easypure™ II or Easypure™ RF water system with Barnstead™ Easypure™ Cartridges and Filters. Choose the cartridge pack according to your feed water and system.
D502126:
  • Includes: Organic/Colloid; two Thermo Scientific™ Ultrapure™ Ion Exchange
  • Type 1 filter
D502124:
  • Includes: Organic/Colloid, Ultrapure Ion Exchange, Organic Free Organic Removal
  • Ultralow filter with Organic Free Cartridge
D502127:
  • Includes: 09-034-18 Organic/Ion Exchange, two 09-050-46 Ultrapure Ion Exchange
  • Type 1 filter
D502125:
  • Includes: 09-034-18 Organic/Ion Exchange, 09-050-46 Ultrapure Ion Exchange, 09-034-16 Organic Free Organic Removal
  • Ultralow filter with Organic Free Cartridge
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Feedwater Source Reverse Osmosis, Distilled
For Use With (Equipment) Easypure II UV, Easypure II UV/UF
Type Cartridge Pack, RO or Distilled Feed