Learn More
Thermo Scientific™ Barnstead™ Easypure™ RoDi Accessories and Consumables
Catalog No. p-4531500
Change view
Click to view available options
Description:
Cartridge Kit
Pre-filtration Accessory Kit
Replacement Ultraviolet Lamp
Start-up Kit
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
13-935-84 Pre-filtration Accessory Kit
13-935-81 Start-up Kit
13-935-82 Cartridge Kit
09-047-81 Replacement Ultraviolet Lamp
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 13-935-84 Supplier Thermo Scientific™ Supplier No. AY1332X2
Only null left
Add to Cart
Add to Cart
Customize your water purification system with the Thermo Scientific™ Barnstead™ Easypure™ RoDi Accessories and Consumables that best suit your application.
Customize your water purification system with the Thermo Scientific™ Barnstead™ Easypure™ RoDi Accessories and Consumables that best suit your application. Cartridge kit includes pre-filter, ultrapure mixed bed cartridge and Easypure high purity/low TOC cartridge. Other consumables include 0.2 micron final filter, ultraviolet lamp, Thermo Scientific™ Ventgard™ reservoir cap, reverse osmosis membrane, and set of three empty cartridges for cleaning.
TRUSTED_SUSTAINABILITY
Specifications
Description Pre-filtration Accessory Kit
For Use With (Equipment) Barnstead Easypure RoDi
Type Pre-filtration Accessory Kit
Quantity Each