Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Barnstead™ Final Filter
GSA_VA

Catalog No. 09034145
Change view
Click to view available options
Pore Size:
0.2 μm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size
09-034-145 0.2 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 09-034-145 Supplier Thermo Scientific™ Supplier No. D3750
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Use the Thermo Scientific™ Barnstead™ Final Filter on select models of Barnstead pure and ultrapure water systems.

Use the Thermo Scientific™ Barnstead™ Final Filter on select models of Barnstead pure and ultrapure water systems. Compatible with Thermo Scientific™ B-Pure™, Nanopure™, Easypure™ II, Easypure™ II RF, Easypure™ RoDi, E-Pure™ (3,4 holder) and TII systems.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description 0.2µm Final Filter
Pore Size 0.2 μm
For Use With (Equipment) B-Pure, Nanopure, Easypure II, Easypure II RF, Easypure RoDi, E-Pure (3,4 holder), and TII systems
Type Final Filter
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.