Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Barnstead™ Hose Nipple Cartridges

Catalog No. 09050264 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
1/2 Size Mixed-Bed with Oxygen Removal
2/3 Size High Capacity
Full Size High Capacity
Full Size Organic Removal
Full Size Pretreatment and Scale Eliminator
Full Size Ultrapure
Full Size Ultrapure with Organic Removal
Hose Nipple Cartridge Holder
Hose nipple cartridge, Full Size Cation Removal
Resin Type:
Activated Carbon
Cation Exchange
Mixed-bed Ion Exchange with Activated Carbon
Mixed-bed and Strong Cation Exchange, Activated Charcoal
Two Bed Ion Exchange
Two Bed Ion Exchange with Strong Cation Exchange
Ultrapure Ion Exchange
Ultrapure Mixed Bed Ion Exchange
Ultrapure Mixed Bed with Strong Anion Exchange
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Resin Type
09-050-264 1/2 Size Mixed-Bed with Oxygen Removal Ultrapure Mixed Bed with Strong Anion Exchange
09-035-20 Full Size High Capacity Two Bed Ion Exchange
09-035-25 Full Size Ultrapure Ultrapure Mixed Bed Ion Exchange
09-035-22 Full Size Organic Removal Activated Carbon
09-034-12 Hose nipple cartridge, Full Size Cation Removal Cation Exchange
09-034-14 Full Size Ultrapure Ultrapure Ion Exchange
09-035-32 Full Size Pretreatment and Scale Eliminator Mixed-bed and Strong Cation Exchange, Activated Charcoal
09-035-30 Full Size Ultrapure with Organic Removal Mixed-bed Ion Exchange with Activated Carbon
09-035-28 2/3 Size High Capacity Two Bed Ion Exchange with Strong Cation Exchange
09-035-50 Hose Nipple Cartridge Holder -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 09-050-264 Supplier Thermo Scientific™ Supplier No. D8822
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Purify water up to 75 L/hr using these hose nipple cartridges, ideal for routine lab work including pretreatment.

Purify water volumes up to 75L/hr using cost-efficient Thermo Scientific™ Barnstead™ Hose Nipple Cartridges. Ideal for softening, deionization, and organic and chlorine removal applications, these cartridges are for use with the simple cartridge holder that easily mounts to the wall. Designed for non-pressure applications, these cartridges include 3/8 in. barb hose connections on each end. They provide the ability to see when resin is exhausted in specific cartridges by a color change indicator. Constructed of 100% virgin polypropylene cartridge, the system is customizable and an ideal option for routine laboratory work.

  • Ideal for small volume applications
  • Same superior quality resins used in B-Pure cartridges
  • Constructed of 100% virgin polypropylene cartridge
  • Includes 3/8″ barb hose connections on each end D8822, D8950, and D8951 cartridges include tapered straight nipple for 3/8″ ID tubing
  • Color change indicator that identifies when resin is exhausted in specific cartridges
TRUSTED_SUSTAINABILITY

Specifications

Description 1/2 Size Mixed-Bed with Oxygen Removal
For Use With (Equipment) Barnstead Water Purification System
Particle Retention 280 grains/4.4 g
Type Cartridge, Hose Nipple
Flow Rate Up to 75 L/hr.
For Use With (Application) Ultrapure Mixed-Bed with Strong Anion Exchange
Resin Type Ultrapure Mixed Bed with Strong Anion Exchange
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.