Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Barnstead™ LabTower™ TII Water Purification System
GSA_VA

Catalog No. 10451088 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
Barnstead LabTower TII (UV)
Barnstead LabTower TII 60
Permeate performance:
40 L/hr. at 15°C
60 L/hr. at 15°C
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Permeate performance
10-451-088 Barnstead LabTower TII (UV) 40 L/hr. at 15°C
10-451-219 Barnstead LabTower TII 60 60 L/hr. at 15°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Catalog No. 10-451-088 Supplier Thermo Scientific™ Supplier No. 50132141
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Convert tap water into high-purity water with the Thermo Scientific™ Barnstead™ LabTower™ TII Water Purification System, a complete solution integrating purification and tank.

Convert tap water into high-purity water with the Thermo Scientific™ Barnstead™ LabTower™ TII Water Purification System, a complete solution integrating purification and tank. This high-tech system is designed to continuously recirculate purified reservoir water to preserve water quality even during periods of inactivity. Conserve bench space with this stand-alone design which includes an integrated 100L reservoir. Castors allow for easy relocation within the lab.

Consistent water quality:

  • Type 2 laboratory water meets ASTM Type II, CSLS-CLRW and ISO 3696 international standards.
  • The LabTower is designed to continuously recirculate purified reservoir water to preserve water quality even during periods of inactivity.

Compact mobile design:

  • Three system options are available with permeate flows of 20, 40 or 60L/hr.
  • Stand-alone design with integrated 100L reservoir takes up zero bench space.
  • Castors allow easy relocation.
  • Systems can be upgraded later to accommodate growing water demands.

Two ways to dispense water:

  • Dispense directly from the system via the dispensing valve for calibration solutions, reagent preparation, filling of containers, general rinsing purposes, etc.
  • Dispense from the reservoir to supply autoclaves, dishwashers, ultrapure water systems, etc.

Integrated 100L reservoir:

  • Up to 100L of high purity water is ready for dispensing when needed.
  • High purity polypropylene reservoir is opaque to light.
  • Conical bottom allows complete emptying for efficient cleaning and disinfection.
  • Adjustable setting for reservoir volume can be programmed for times when demand is low, allowing you to only store water that you can use.

Clear information on system display:

  • The large four-line display is code-protected to prevent accidental changes in system settings.
  • Illuminated and easily readable, the display provides information on resistivity or conductivity and operating mode status, such as “roduction”, “stand-by” or sanitization and volume of the reservoir in percent.

GLP-compliant documentation:

  • LabTower TII has been developed to meet or exceed GLP requirements.
  • Recorded and traceable data can be obtained by print-out via the RS-232 interface and accessory printer.
  • Highly qualified and precise measurement of the conductivity is ensured by the cell constant of 0.01 cm-1 to an accuracy of ±0.1°C.

Pretreatment:

  • 5μm prefilter and activated carbon in the combi-filter cartridge protect the RO membrane from free chlorine and particles.
  • Hardness stabilizing cartridge protects the reverse osmosis stage from hard water.

Reverse osmosis and reservoir:

  • Pretreated water is pressure-forced through the permeable reverse osmosis module removing 97- 99% of all inorganic ions and 99% of dissolved organic substances, as well as microorganisms and particles.
  • The built-in 100L reservoir has a conical bottom outlet to optimize cleaning and sanitization.

Ion exchange and UV:

  • RO membrane removes up to 99% of impurities. All remaining ions are removed by the low TOC, high-purity resins in the ion exchange cartridge.
  • Irradiation with UV light (TII units with UV option only) eliminates any bacteria or germs that are present.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Bacterial retention 99%
Conductivity 0.1 to 1.0μS/cm
Description Barnstead LabTower TII (UV)
Bacterial Content <1 CFU/mL
Feedwater Connector 3/4 in. NPT
Feed Water Monitoring No
Feedwater Source Tap
Flow Rate LabTower TII (UV) 40 with 3 L/min. pump
Product Water Type Type 2
Resistivity 10 to 1 Mohm.cm
TOC Monitor with UV Intensity Monitoring No
Feedwater Pressure 1 to 6 bar
Operating Pressure 2 to 6 bar
Particles >0.2µM/mL <1
Permeate performance 40 L/hr. at 15°C
Temperature Range (Metric) 2°C to 35°C
Width (Metric) 450 mm
Height (Metric) 1500 mm
Depth (Metric) 580 mm
Net Weight (Metric) 66 kg
Electrical Requirements 100/240 V 50/60 Hz
Voltage 100/240 V
Frequency 50/60 Hz
Product Type Water Purification System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.