Learn More
Thermo Scientific™ Barnstead™ MegaPure™ ACS
Catalog No. 0906978 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Adapter Kit
Automatic Collection System
Wall-mount Bracket
For Use With (Equipment):
Automatic Collection System
Mega-Pure Glass Still
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type For Use With (Equipment)
09-069-78 Wall-mount Bracket Automatic Collection System
09-069-76 Adapter Kit Mega-Pure Glass Still
09-069-72 Automatic Collection System Mega-Pure Glass Still
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 09-069-78 Supplier Thermo Scientific™ Supplier No. 400634
Add to Cart
Add to Cart
Control the operation of your Thermo Scientific™ Barnstead™ Mega-Pure™ Glass Still with this automatic collection system, which collects up to 45L of water.
Control the operation of your glass still with the Thermo Scientific™ Barnstead™ MegaPure™ ACS (Automatic Collection System), which collects up to 45L of water.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Automatic Collection System
Type Wall-mount Bracket
Quantity Each