Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Barnstead™ MegaPure™ Glass Stills

Catalog No. 09081
Change view
Click to view available options
Output Flow Rate:
0.4 gal./hr.
0.9 gal./hr.
1.6 gal./hr.
Voltage:
120 V
208 V
240 V
Model:
MP-1
MP-1 w/6L bottle
MP-3A
MP-6A
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Output Flow Rate Voltage Model
09-081 0.4 gal./hr. 120 V MP-1
09-080 0.4 gal./hr. 120 V MP-1 w/6L bottle
09-085 0.9 gal./hr. 240 V MP-3A
09-089 1.6 gal./hr. 240 V MP-6A
09-085-1 0.9 gal./hr. 208 V MP-3A
09-088 1.6 gal./hr. 208 V MP-6A
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 09-081 Supplier Thermo Scientific™ Supplier No. A440266
Add to Cart
Add to Cart
Request a Quote

Obtain high purity water from distillation using these glass stills, constructed of non-leaching components.

Obtain high purity water from distillation using Thermo Scientific™ Barnstead™ Mega-Pure™ Glass Stills, constructed of non-leaching components. These glass stills effectively remove inorganic solids, organics with boiling points higher than water, bacteria and pyrogens. Available in five production capacities, units are easily wall- or bench-mounted. Maximum purity is provided from the vertical condenser design. Gain easy access for cleaning with the quick release cover.

Features:
  • Contact with only glass, quartz and PTFE components ensures ultimate purity and eliminates cross-contamination.
  • Vertical condenser design provides maximum purity.
  • High temperature cutoff shuts down the still if the temperature is too high, preventing heating element burnout.
  • Quick-release cover provides easy cleaning access.
  • Connection to optional Automatic Connection System (ACS) assures a 24-hour automatic system.
  • Stills include feedwater solenoid valve for automatic operation.
  • Stills produce water with a resistivity of 1 to 10MΩ-cm, depending on feedwater quality.

MP-1:

  • Compact, easy-to-use 1L/hr. glass still
  • Available with or without a storage bottle

MP-3A, MP-6A and MP-11A:

  • For larger volume demands with storage and options (3.4 to 13L/hr.)

MP-12A:

  • Features built-in deionizer for pretreated feed to boiler
  • Distillate cooler allows product water to be used as it is produced, cooling it from 85°C (185°F) to about 55°C (131°F) for ease of handling
  • No need to wait for cooling

Ordering Information:

Optional accessories include single or dual cartridge deionizers (D440046 or D440066), adapter kits (440376 or 440375), and replacement cartridges (D400377, D400499 or D440265). Storage options include bottles (413964, 410535, 413934, 410164), or Automated Collection System (B440704). All sold separately; contact your sales representative for details.

This product is not available in all regions.

Includes:

Feed water solenoid valve for automatic operation

Requires:

Water supply and storage reservoir

Dual Solenoid required if using pretreated (DI, distilled, RO) feedwater:

  • 440236 - for use with MP-1 (240V), MP-3A, MP-6A, and MP-11A
  • RY798X2A - for use with MP-1 (120V)

Deionization Cartridges required for MP-12A:

  • D440046 - Single Cartridge Deionizer (D1)
  • D440066 - Dual Cartridge Deionizer (D2)

All accessories sold separately

Recommended for:

Buffer/reagent prep, glassware washing, dishwasher feed

TRUSTED_SUSTAINABILITY

Specifications

Type Glass Still
Frequency 50/60 Hz
Voltage 120 V
Amperage 9 A
Flow Rate 0.4 gal./hr.
Electrical Requirements 120 V 50/60 Hz
Inlet Water Temperature (Metric) 4°C to 37°C
Inlet Pressure 20 to 100 psi
Vent Temperature 85°C to 96.7°C
Output Flow Rate 0.4 gal./hr.
Model MP-1
Auto Drain No
Depth (English) 9.75 in.
Depth (Metric) 24.8 cm
Height (English) 34 in.
Height (Metric) 86.4 cm
Width (English) 18 in.
Width (Metric) 45.7 cm
Show More Show Less

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.