Learn More
Thermo Scientific™ Barnstead™ Nanopure™ Accessories and Consumables
GSA_VA
Catalog No. 09050284 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Deionized Water Feed (Low Organics, Type I Water)
Deionized Water Feed (Ultralow Organics, Type I Water)
R/O and Distilled Feed (Low Organics, Type I Water)
R/O and Distilled Feed (Ultralow Organics, Type I Water)
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
09-050-284 R/O and Distilled Feed (Ultralow Organics, Type I Water)
09-050-283 Deionized Water Feed (Ultralow Organics, Type I Water)
09-035-13 R/O and Distilled Feed (Low Organics, Type I Water)
09-050-282 Deionized Water Feed (Low Organics, Type I Water)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 09-050-284 Supplier Thermo Scientific™ Supplier No. D50280
  • $2,980.00
    $1,861.65 / Each of 1
    Save $1,118.35
    38% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Optimize your water purification system with Thermo Scientific™ Barnstead™ Nanopure™ Accessories and Consumables.
Optimize your water purification system with Thermo Scientific™ Barnstead™ Nanopure™ Accessories and Consumables. Choose the cartridge pack according to your feed water and system. Cartridge packs ship with 0.2 micron final filter. Other products include UV lamp and low water level pump protector.
  • Simple, one-connection installation and removal
Includes: One 0.2μm absolute final filter
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Nanopure Water Purifications System
Type R/O and Distilled Feed (Ultralow Organics, Type I Water)