Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Barnstead™ Pretreatment for Point of Use Water Purification Systems

Catalog No. 11676673 Shop All Thermo Scientific Products
Change view
Click to view available options
Cartridge Type:
Pretreatment Cartridge
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cartridge Type
11-676-673 Pretreatment Cartridge
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-676-673 Supplier Thermo Scientific™ Supplier No. 50157886
  • $2,300.00
    $1,764.65 / Each of 1

    Save $535.35

    23% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Barnstead Pretreatment for Point of Use Water Purification Systems

Barnstead Pretreatment contains PreFilter, Hardness Stabilizer and Carbon Filter. The unit contains pressure regulator and pressure gauges. Space saving design comes with Wall Mount capability.

  • Use pretreatment when feed water quality doesn't match the requirements for your system
  • Contains pre-filter, hardness stabilizer and carbon filter
TRUSTED_SUSTAINABILITY

Specifications

Cartridge Type Pretreatment Cartridge
Description Pretreatment includes Particle Filter, Carbon Filter and Hardness Stabilizer Catridges ( D502113, D502114 and D502115 )
For Use With (Equipment) Smart2Pure Pro
Includes Pressure Regulator, Pressure Gauges, Three Cartridges and Wall Mount
Dimensions (L x W x H) 14.5 in x 13.3 in x 20.0 in.
Weight (English) 24.5 lb.
Weight (Metric) 11.4 kg
Product Type Water Purification System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.