Learn More
MSC BARRACUDA C/R GLOVE
Catalog No. 5000675978
Encompass
PIP Barracuda Cut Force Hi Vis Polykor Nitrile Coated Foam Grip Gloves are lightweight, offering excellent dexterity, tactile sensitivity, and cut resistance. They are ideal for applications like electronics, glass cutting, sheet metal handling, parts assembly, and construction.
  • Polykor blended shell provides excellent dexterity, tactile sensitivity, and cut resistance.
  • Seamless construction enhances comfort and breathability.
  • Foam Nitrile coated palm and fingers offer good grip and abrasion resistance, compatible with light oils.
  • Knit wrist helps prevent dirt and debris from entering the glove.
  • Hi-Vis coloring for high visibility.
  • Color coded hems for easy size identification.
  • Washable, resistant to chemicals, water, and UV light.
Catalog No. 50-006-75978 Supplier MSC Supplier No. 96262688
PIP Barracuda Cut Force Hi Vis Polykor Nitrile Coated Foam Grip Gloves are lightweight, offering excellent dexterity, tactile sensitivity, and cut resistance. They are ideal for applications like electronics, glass cutting, sheet metal handling, parts assembly, and construction.
  • Polykor blended shell provides excellent dexterity, tactile sensitivity, and cut resistance.
  • Seamless construction enhances comfort and breathability.
  • Foam Nitrile coated palm and fingers offer good grip and abrasion resistance, compatible with light oils.
  • Knit wrist helps prevent dirt and debris from entering the glove.
  • Hi-Vis coloring for high visibility.
  • Color coded hems for easy size identification.
  • Washable, resistant to chemicals, water, and UV light.