Learn More
Med Vet International Miltex Barraquer Eye Speculum, 1.75in., 14mm Wide Blades, Non-Magnetic
Catalog No. 502829089
Encompass_Preferred
The Miltex 18-35 Barraquer eye speculum is made of OR grade German stainless steel and measures 1.75" in length. It has large blades, 14mm wide, and it is nonmagnetic.
Catalog No. 50-282-9089 Supplier Med Vet International Supplier No. MIL1835
The Miltex 18-35 Barraquer eye speculum is made of OR grade German stainless steel and measures 1.75" in length. It has large blades, 14mm wide, and it is nonmagnetic.