Learn More
Electron Microscopy Sciences Aluminium Barrel Microtome Chuck with Spigot, 22 mm diameter
SDP
Catalog No. 501930528
Encompass_Preferred
Aluminum Microtome Chucks with grooved surfaces for good specimen adhesion. Chucks have a 9.4 mm spigot.
Diameter: 22 mm
Catalog No. 50-193-0528 Supplier Electron Microscopy Sciences Supplier No. EMS067
Aluminum Microtome Chucks with grooved surfaces for good specimen adhesion. Chucks have a 9.4 mm spigot.
Diameter: 22 mm