Learn More
BURKLE INC PTFE barrel pump Ultrapure with d. hose/stopc., 23.62 in. (60 cm)
Catalog No. 5000026005
Encompass_Preferred
Due to the high chemical resistance of the wetted materials PTFE and FEP, this drum pump can also be used to fill liquids that corrode conventional materials such as PP or PVC (with the exception of elemental fluorine).
The drum pump is also suitable for filling high-purity liquids.
The stopcock prevents hazardous vapors from escaping and liquids from dripping after filling.
Catalog No. 50-000-26005 Supplier BURKLE INC Supplier No. 56060601
Due to the high chemical resistance of the wetted materials PTFE and FEP, this drum pump can also be used to fill liquids that corrode conventional materials such as PP or PVC (with the exception of elemental fluorine).
The drum pump is also suitable for filling high-purity liquids.
The stopcock prevents hazardous vapors from escaping and liquids from dripping after filling.