Learn More
BURKLE INC PP barrel pump with discharge hose/stopcock, 49.21 in. (125 cm)
Catalog No. 5000025636
Encompass_Preferred
Barrel pumps made of polypropylene allow a fast transfer of liquids from barrels, canisters and tanks.
The low-mounted non-return valve of the barrel pump and the immersion depth that can be steplessly adjusted using the barrel screw joints ensure good removal of residual liquid. The barrel screw joint fastens the barrel's pump securely to the barrel. Barrel screw joints and thread adapters for commercially available containers are available as optional accessories.
Catalog No. 50-000-25636 Supplier BURKLE INC Supplier No. 56001252
Barrel pumps made of polypropylene allow a fast transfer of liquids from barrels, canisters and tanks.
The low-mounted non-return valve of the barrel pump and the immersion depth that can be steplessly adjusted using the barrel screw joints ensure good removal of residual liquid. The barrel screw joint fastens the barrel's pump securely to the barrel. Barrel screw joints and thread adapters for commercially available containers are available as optional accessories.