Learn More
BURKLE INC PP barrel pump with discharge tube, 59.06 in. (150 cm), 10.14 oz. (300 ml)/stroke
Catalog No. 5000025711
Encompass_Preferred
Barrel pumps made of polypropylene allow a fast transfer of liquids from barrels, canisters and tanks.
The low-mounted non-return valve of the barrel pump and the immersion depth that can be steplessly adjusted using the barrel screw joints ensure good removal of residual liquid. The barrel screw joint fastens the barrel's pump securely to the barrel. Barrel screw joints and thread adapters for commercially available containers are available as optional accessories.
Catalog No. 50-000-25711 Supplier BURKLE INC Supplier No. 56001501
Barrel pumps made of polypropylene allow a fast transfer of liquids from barrels, canisters and tanks.
The low-mounted non-return valve of the barrel pump and the immersion depth that can be steplessly adjusted using the barrel screw joints ensure good removal of residual liquid. The barrel screw joint fastens the barrel's pump securely to the barrel. Barrel screw joints and thread adapters for commercially available containers are available as optional accessories.