Learn More
BURKLE INC Stainless steel barrel pump withd. hose/stopcock, 14.17 in. (36 cm)
Catalog No. 5000025900
Encompass_Preferred
Stainless steel drum pumps are powerful, versatile piston pumps for transferring flammable or aggressive liquids and chemicals.
Their electrical conductivity makes them ideal for highly flammable liquids. Developed for zones 0 and 1 (explosion groups II A and II B), they are DEKRA-tested. Pumps must be grounded when used with flammable media.
PTFE seals guarantee high chemical resistance and excellent sliding during pumping.
These pumps fit various drums and containers (60 to 1000 l, up to 36 cm depth), making them true all-rounders in workshops and production.
Compared to conventional solvent pumps, they offer excellent value.
Made of corrosion-resistant stainless steel (1.4301), they ensure long service life, especially in humid or aggressive environments.
Catalog No. 50-000-25900 Supplier BURKLE INC Supplier No. 56010401
Stainless steel drum pumps are powerful, versatile piston pumps for transferring flammable or aggressive liquids and chemicals.
Their electrical conductivity makes them ideal for highly flammable liquids. Developed for zones 0 and 1 (explosion groups II A and II B), they are DEKRA-tested. Pumps must be grounded when used with flammable media.
PTFE seals guarantee high chemical resistance and excellent sliding during pumping.
These pumps fit various drums and containers (60 to 1000 l, up to 36 cm depth), making them true all-rounders in workshops and production.
Compared to conventional solvent pumps, they offer excellent value.
Made of corrosion-resistant stainless steel (1.4301), they ensure long service life, especially in humid or aggressive environments.