Learn More
BURKLE INC PP barrel screw joint, G 2", outer
Catalog No. 5000025543
Encompass_Preferred
Barrel screw joint made of PP for secure connection in the barrel in three different thread models. For drawing off liquids that are not flammable.
The container connectors make decanting even easier and safer with their more secure connection to barrels. The connectors secure the pump in the barrel, enable a vertical position change and allow the required air exchange.
Catalog No. 50-000-25543 Supplier BURKLE INC Supplier No. 56003130
Barrel screw joint made of PP for secure connection in the barrel in three different thread models. For drawing off liquids that are not flammable.
The container connectors make decanting even easier and safer with their more secure connection to barrels. The connectors secure the pump in the barrel, enable a vertical position change and allow the required air exchange.