Learn More
Med Vet International EasyTouch Luer Lock Syringe Barrels, Insulin, 1ml, 100/Box
Catalog No. 502559687
Encompass_Preferred
A clear barrel is a better barrel! EasyTouch Barrels are designed and manufactured to be ultra clear with well defined markings. Designed with minimal dead space. Ultra-clear Barrel Well-defined Markings Smooth Surface for Low Friction Dispensing Latex Free
Catalog No. 50-255-9687 Supplier Med Vet International Supplier No. 802015
A clear barrel is a better barrel! EasyTouch Barrels are designed and manufactured to be ultra clear with well defined markings. Designed with minimal dead space. Ultra-clear Barrel Well-defined Markings Smooth Surface for Low Friction Dispensing Latex Free