Learn More
Med Vet International EasyTouch Luer Lock Tuberculin Syringe Barrels, TB, 10ml, 50/Box
Catalog No. 502559684
Encompass_Preferred
A clear barrel is a better barrel! EasyTouch Barrels are designed and manufactured to be ultra clear with well defined markings. Designed with minimal dead space. Ultra-clear Barrel Well-defined Markings Smooth Surface for Low Friction Dispensing Latex Free
Catalog No. 50-255-9684 Supplier Med Vet International Supplier No. 802100
A clear barrel is a better barrel! EasyTouch Barrels are designed and manufactured to be ultra clear with well defined markings. Designed with minimal dead space. Ultra-clear Barrel Well-defined Markings Smooth Surface for Low Friction Dispensing Latex Free