Learn More
Med Vet International EasyTouch Luer Lock Syringe Barrels, TB, 5ml, 100/Box, 5 Boxes
Supplier:  Med Vet International 802050X5
Encompass_Preferred
A clear barrel is a better barrel! EasyTouch Barrels are designed and manufactured to be ultra clear with well defined markings. Designed with minimal dead space. Ultra-clear Barrel Well-defined Markings Smooth Surface for Low Friction Dispensing Latex Free
Catalog No. 50-209-1641
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.