Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform™ Barricade Light

Catalog No. 19169601 Shop All AccuformNMC Products
Change view
Click to view available options
Product Type:
Incandescent Barricade Light
Solar Barricade Light
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19-169-601 Solar Barricade Light
19-169-600 Incandescent Barricade Light
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-169-601 Supplier Accuform™ Supplier No. FBL639
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Accuform™ Barricade Lights add visibility and draw attention to warn of hazards situations in low-light conditions

Incandescent:

  • Durable polycarbonate 7 in. amber lens with plastic yellow battery-case base
  • Incandescent light can be set to steady-on or flashing
  • Photocell turns on automatically in dark conditions
  • Concealed on-off switch with bolt and set-in nut included for easy mounting
  • Requires two, 6-volt batteries (not included)

Solar:

  • Eliminate the maintenance, replacement, and disposal costs of batteries and bulbs
  • Solar photovoltaic cells located on top of the 7 in. diameter x 3 ¾ in. high-impact, polycarbonate lens
  • Durable polypropylene case has universal mount bracket and 360° swivel
  • On/off and steady-on/flash switches secured inside small holes
  • High-intensity LED with steady-on brightness of 2000 MCD, or flashing brightness of 4000 MCD with flash rate of 60 per minute
  • Batteries operate up to three nights on single charge
  • Photocell activates device when ambient light falls below 215 lux

Specifications

Product Type Solar Barricade Light
Color Amber-Yellow
Type Solar Barricade Light
Dimensions (L x W) 7 x 3.75 in.
Material Polycarbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.