Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform™ Barrier Cone Bar

Catalog No. 19169597 Shop All AccuformNMC Products
Change view
Click to view available options
Color:
Orange, White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-169-597 Orange, White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-169-597 Supplier Accuform™ Supplier No. FBC604
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Creates a quick, temporary barrier when attached to traffic cones to section-off hazards and danger zones.

  • Portable, expandable barrier
  • Ring-ends fit over traffic cones (sold separately)
  • Reflective stripes for visibility in low-light conditions
  • Expands up to 6 ½ ft.
  • Collapses to 4 ft. for storage and transport

Specifications

For Use With (Equipment) Traffic Cones
For Use With (Application) Temporary Barriers
Length (English) 6.5 ft. (Max)
Product Type Expandable Bar
Color Orange, White
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.