Learn More
Med Vet International Dynarex Dental Barrier Film, 4" X 6", Clear, 9600/Case
Supplier:  Med Vet International 4488
Encompass_Preferred
Dynarex Dental Barrier Film is designed provide a protective barrier for a wide range of handheld instruments, including dental supplies, medical tools, and other essential equipment. Ideal for dental offices, clinics, and medical facilities, the Barrier Film adheres securely to most surfaces without leaving a sticky residue behind. Each roll of Barrier Film comes on a plastic spool and is divided into 1200 perforated sheets for easy tearing. The Film is packed in a dispenser box with convenient cut-out opening for quick and efficient dispensing. Available in a choice of blue or clear.  Dynarex Dental Barrier Film is a high-quality plastic wrap designed to help prevent cross-contamination when using handheld instruments and to help protect difficult-to-clean surfaces. The Dental Barrier Film with self-adhesive backing is divided into perforated sheets for easy tearing and comes in a convenient, slotted dispenser box for quick access and efficient dispensing. 
Catalog No. 50-283-1595