Learn More
Andwin Scientific BARRIER TIP 1000 UL 3072/CS
SDP
Catalog No. NC0515691
Encompass_Preferred
The Neptune BT Barrier Pipet Tips are constructed from virgin polypropylene. This product hinders aerosol contamination and offer excellent consistency. It features a thin wall design that gives an optimal seal for both single and multichannel pipettors. The tips have a universal fit and low retention. These have a volume capacity of 1000 L and S3 Polymer Tips that remove sample hold up and guarantee a more precise drawing and dispensing of samples. This product is sold in a case of 3072 tips.
Catalog No. NC0515691 Supplier Andwin Scientific Supplier No. 89217468
The Neptune BT Barrier Pipet Tips are constructed from virgin polypropylene. This product hinders aerosol contamination and offer excellent consistency. It features a thin wall design that gives an optimal seal for both single and multichannel pipettors. The tips have a universal fit and low retention. These have a volume capacity of 1000 L and S3 Polymer Tips that remove sample hold up and guarantee a more precise drawing and dispensing of samples. This product is sold in a case of 3072 tips.