Learn More
Andwin Scientific BARRIER TIPS 1000UL 3072CS
SDP
Catalog No. NC0762898
Encompass_Preferred
The Neptune BT Barrier Pipet Tips are constructed from virgin polypropylene. This product hinders aerosol contamination and offer excellent consistency. It features a thin wall design that gives an optimal seal for both single and multichannel pipettors. The tips have a universal fit and low retention. These have a volume capacity of 1000 L. This product is sold in a case of 3072 tips.
Catalog No. NC0762898 Supplier Andwin Scientific Supplier No. 89217470CS
The Neptune BT Barrier Pipet Tips are constructed from virgin polypropylene. This product hinders aerosol contamination and offer excellent consistency. It features a thin wall design that gives an optimal seal for both single and multichannel pipettors. The tips have a universal fit and low retention. These have a volume capacity of 1000 L. This product is sold in a case of 3072 tips.