Learn More
Hach Company BART Test for Denitrifying Bacteria, pk/9
Catalog No. 501996375 Shop All Hach Company Products
Encompass
BART biodetectors are excellent diagnostic tools to help identify the presence and activity of various bacteria. Denitrifying bacteria indicate the decomposition of waste organic nitrogenous materials. These bacteria reduce nitrate to nitrite and some continue nitrification to gaseous nitrogen (complete denitrification). In water, aggressive denitrifiers can indicate high concentrations of nitrates, and that the sample is probably anaerobic and relatively rich in organic matter. The presence of denitrifying bacteria can indicate that the water has been polluted by nitrogen-rich organics from sources such as compromised septic tanks, sewage systems, industrial and hazardous waste sites. If highly aggressive bacteria are detected, the water should be tested for the presence of coliform bacteria.
Catalog No. 50-199-6375 Supplier Hach Company Supplier No. 2619309
BART biodetectors are excellent diagnostic tools to help identify the presence and activity of various bacteria. Denitrifying bacteria indicate the decomposition of waste organic nitrogenous materials. These bacteria reduce nitrate to nitrite and some continue nitrification to gaseous nitrogen (complete denitrification). In water, aggressive denitrifiers can indicate high concentrations of nitrates, and that the sample is probably anaerobic and relatively rich in organic matter. The presence of denitrifying bacteria can indicate that the water has been polluted by nitrogen-rich organics from sources such as compromised septic tanks, sewage systems, industrial and hazardous waste sites. If highly aggressive bacteria are detected, the water should be tested for the presence of coliform bacteria.