Learn More
Hach Company BART Test for Fluorescing Pseudomonas, pk/9
Supplier:  Hach Company 2432609
Encompass
BART biodetectors are excellent diagnostic tools to help identify the presence and aggressivity of various bacteria. Pseudomonad bacteria are often present in waters that contain oxygen and are rich in organic pollutants (e.g., gasoline, jet fuel, solvents). The presence of pseudomonad bacteria may indicate that aerobic biodegradation is occurring and biofouling may also be happening within the system being tested. Some pseudomonad bacteria that produce the fluorescent pigments (pigments that glow in ultraviolet light) may be a hygiene risk. The faster that clouding and fluorescence happens, the more aggressive are the pseudomonad bacteria.Pseudomonad bacteria can cause a range of problems in water, including slime formations, turbidity, taste and odor, corrosion, biodegradatrion, and hygiene risks. In recreational waters (such as swimming pools, hot tubs, restricted natural bathing sites), the presence of aggressive fluorescent pseudomonads can cause various infections.
Catalog No. 50-199-5981