Learn More
Hach Company BART Test for Heterotropic Aerobic Bacteria, pk/9
Supplier:  Hach Company 2490409
Encompass
BART biodetectors are excellent diagnostic tools to help identify the presence and activity of various bacteria. Heterotrophic Aerobic Bacteria (HAB) are one of the most important agents in biofouling, causing corrosion, clogging, fouling of the water, and increased hygiene risks.
Catalog No. 50-199-6069