Learn More
Hach Company BART Test for Nitrifying Bacteria, pk/7
Catalog No. 501996376 Shop All Hach Company Products
Encompass
BART biodetectors are excellent diagnostic tools to help identify the presence and activity of various bacteria. Nitrifying bacteria recycle organic nitrogenous materials from ammonium (the endpoint for the decomposition of proteins) to nitrates. In water, aggressive nitrifiers can produce high concentrations of nitrates. Nitrates in water can be a potential health risk. Aggressive nitrifying bacteria in waters may indicate the latter stages of aerobic degradation of nitrogen-rich organic matter. This can indicate that the water may have been polluted by nitrogen-rich organics from sources such as compromised septic tanks, sewage systems, industrial and hazardous waste sites and is undergoing an aerobic form of degradation.
Catalog No. 50-199-6376 Supplier Hach Company Supplier No. 2619407
BART biodetectors are excellent diagnostic tools to help identify the presence and activity of various bacteria. Nitrifying bacteria recycle organic nitrogenous materials from ammonium (the endpoint for the decomposition of proteins) to nitrates. In water, aggressive nitrifiers can produce high concentrations of nitrates. Nitrates in water can be a potential health risk. Aggressive nitrifying bacteria in waters may indicate the latter stages of aerobic degradation of nitrogen-rich organic matter. This can indicate that the water may have been polluted by nitrogen-rich organics from sources such as compromised septic tanks, sewage systems, industrial and hazardous waste sites and is undergoing an aerobic form of degradation.