Learn More
Hach Company BART Test, Slime-Forming Bacteria, pk/9
Supplier:  Hach Company 2432509
Encompass
BART biodetectors are excellent diagnostic tools to help identify the presence of process disruptors, such as bacteria creating slime layers and limiting disinfection.The SLYM-BARTs can be used as a P/A test capable of indicating to some extent the possible population size and the types of slime-forming organisms present in the water sample. Slime-forming bacteria are able to produce copious amounts of slime without necessarily having to use any iron. Iron bacteria also produce slime, but usually it is thinner and involves the accumulation of various forms of iron.Slime-forming bacteria generally produce the thickest slime formations under aerobic (oxidative) conditions, which develop around the floating ball. Growth may be recognized as a cloudy or gel-like growth, which can be localized or occur throughout the sample. These growths are usually white, grey, yellow, or beige in color and can darken over time.
Catalog No. 50-199-5979