Learn More
Aviva Systems Biology Corp BARX2 Antibody - N-terminal region (ARP38371_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38371_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BARX2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human BARX2. Alternate names: MGC133368, MGC133369ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56626