Learn More
Aviva Systems Biology Corp Barx2 Antibody - N-terminal region (ARP37202_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37202_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Barx2. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 2310006E12Rik, Barx2bApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-592