Learn More
Lonza Walkersville EBM Endothelial Cell Growth Basal Medium, Phenol Red Free, 500 mL
Catalog No. NC9762785
Encompass
EBM Phenol Red Free is an endothelial basal medium sold separately. This can be used with 1 kit of CC-4133 (endothelial cell growth medium SingleQuots Supplements and Growth Factors for use with certain HUVEC cells).Choose from a variety of endothelial cell growth media for an easy cultivation of primary endothelial cells. Every endothelial cell growth medium has its special formulation and is optimized to grow certain cell types. Each type of endothelial cell growth medium is guaranteed to perform when used to grow Clonetics Primary Cells.Lonza guarantees the performance of Clonetics/Poietics Cells only if appropriate Clonetics/Poietics Media and Reagents are used exclusively and the recommended storage and use protocols are followed. Content:1 x Bottle EBM Phenol Free Basal Medium, 500 mL
Catalog No. NC9762785 Supplier Lonza Walkersville Supplier No. CC3129
EBM Phenol Red Free is an endothelial basal medium sold separately. This can be used with 1 kit of CC-4133 (endothelial cell growth medium SingleQuots Supplements and Growth Factors for use with certain HUVEC cells).Choose from a variety of endothelial cell growth media for an easy cultivation of primary endothelial cells. Every endothelial cell growth medium has its special formulation and is optimized to grow certain cell types. Each type of endothelial cell growth medium is guaranteed to perform when used to grow Clonetics Primary Cells.Lonza guarantees the performance of Clonetics/Poietics Cells only if appropriate Clonetics/Poietics Media and Reagents are used exclusively and the recommended storage and use protocols are followed. Content:1 x Bottle EBM Phenol Free Basal Medium, 500 mL