Learn More
Lonza Walkersville REBM Renal Epithelial Cell Growth Basal Medium
Catalog No. NC9411805
Encompass
REBM is a renal epithelial cell growth basal medium sold separately. This can be used with 1 kit of CC-4127 (renal cell growth medium SingleQuots Supplements and Growth Factors). The full REGMBulletKit consisting of basal medium and supplements can be purchased together using the catalog number CC-3190.REGM Renal Epithelial Cell Growth Medium BulleKit is formulated to maintain and expand most renal epithelial cells. REGM contains 0.5% serum.Content:1 x Bottle Basal Medium 500 mL
Catalog No. NC9411805 Supplier Lonza Walkersville Supplier No. CC3191
REBM is a renal epithelial cell growth basal medium sold separately. This can be used with 1 kit of CC-4127 (renal cell growth medium SingleQuots Supplements and Growth Factors). The full REGMBulletKit consisting of basal medium and supplements can be purchased together using the catalog number CC-3190.REGM Renal Epithelial Cell Growth Medium BulleKit is formulated to maintain and expand most renal epithelial cells. REGM contains 0.5% serum.Content:1 x Bottle Basal Medium 500 mL