Learn More
Lonza Walkersville HBM Basal Medium, 500 mL
Catalog No. NC9982883
Encompass
The HBMBasal Medium consists of 500Mls hepatocyte basal medium without supplementation. It is designed to be used only in conjunction with the HCM SingleQuots (CC-4182) . Once combinded together these reagents formulate a highly reproducible maintenance medium for primary hepatocytes from all species. The final medium is serum-free.Benefits:Optimized for Hepatocyte Cell Health, Compatible with all species hepatocytesContent:500 mL
Catalog No. NC9982883 Supplier Lonza Walkersville Supplier No. CC3199
The HBMBasal Medium consists of 500Mls hepatocyte basal medium without supplementation. It is designed to be used only in conjunction with the HCM SingleQuots (CC-4182) . Once combinded together these reagents formulate a highly reproducible maintenance medium for primary hepatocytes from all species. The final medium is serum-free.Benefits:Optimized for Hepatocyte Cell Health, Compatible with all species hepatocytesContent:500 mL