Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Basal Medium Eagle (Modified), with Hanks' Salts and L-Glutamine, without Sodium Carbonate

Catalog No. 091003122 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-100-3122 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-100-3122 Supplier MP Biomedicals™ Supplier No. 091003122
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Basal Medium Eagle (Modified), with Hanks' Salts and L-Glutamine, without Sodium Carbonate is one of the most commonly used synthetic basal medium for supporting the growth of many different mammalian cells.

  • Basal Medium Eagle (BME) is one of the most commonly used of all cell culture media.
  • This medium is the result of numerous studies from the late 1950's analyzing the nutrient requirements and other components essential for the growth of cells in culture.
  • Traditionally, BME has been used in studies to measure the growth response of normal (WI-38) and transformed (HeLa) cells in monolayer culture.
  • BME may be used for a broad variety of cell lines when properly supplemented.
  • It has a pH of 6.5 ± 0.5 (w/o sodium bicarb.), 7.3 ± 0.5 (adjusted with sodium bicarb.)
TRUSTED_SUSTAINABILITY

Specifications

Form Powder
For Use With (Application) Bacteria Cell Culture, Mammalian Cell Culture
Quantity 10 L
Formulation Polypeptone 10.0, Biotryptase 10.0, Heart Peptone 3.0, Corn Starch 1.0, Sodium Chloride 5.0, Agar 13.5, Final pH 7.3 ±0.2
pH 6.5 to 7.3 ±0.5
Preparation Method Suspend 42.5 g of the medium in one liter of distilled or deionized water. Mix well and heat with frequent agitation. Boil for one minute. Dispense and sterilize in an autoclave at 121°C (15 lb. psi) for 15 minutes. Cool to 45 to 50°C and add 5 to 10% sterile defibrinated sheep or rabbit blood if desired.
WARNING: Cancer - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.