Learn More
Chemglass Life Sciences Base Only, 47mm, Fritted
SDP
Supplier:  Chemglass Life Sciences CG142004
Encompass_Preferred
Support base for use with the CG142001 filtration support assembly. Support base has a sealed-in 47mm coarse porosity fritted disc. BASE ONLY.
Catalog No. CG142004