Learn More
Binder Inc Base w casters CB/CBS 170
SDP
Supplier:  Binder Inc 90510028
Encompass_Preferred
Base - the base equipped with casters is used for the secure positioning and leveling of a BINDER CO2 incubator
Catalog No. 90-510-028