Learn More
BINDER™ Base with Casters
SDP
Catalog No. 08111585 Shop All BINDER Inc. Products
Change view
Click to view available options
Material:
Steel
Catalog No. Material
08-111-585 Steel
1 options
Catalog No. 08-111-585 Supplier BINDER™ Supplier No. 90510034
Add to Cart
Add to Cart
For Use With: M 240
  • Manufactured from durable materials
  • For a variety of models
Specifications
Description Base - the base equipped with casters is used for the secure positioning and leveling of a BINDER CO2 incubator
For Use With (Equipment) BINDER Chambers M 240
Material Steel
Type Base
Includes Casters