Learn More
Electron Microscopy Sciences Embedding Cassette Disposable Base Mold, 30 x 24 mm, 5 mm height
SDP
Catalog No. 501928633
Encompass_Preferred
Clear plastic. Eliminates use of cleaning solvents and release compounds.
Footprint: 30 x 24 mm
Height: 5 mm
Catalog No. 50-192-8633 Supplier Electron Microscopy Sciences Supplier No. 7091503
Clear plastic. Eliminates use of cleaning solvents and release compounds.
Footprint: 30 x 24 mm
Height: 5 mm