Learn More
Electron Microscopy Sciences Stainless steel for optimal thermal conductivity and polished for easy paraffin block removal. Corners are rounded for good paraffin ribboning. Use for all applications in specimen embedding with Tissue-Tek Embedding Rings.
SDP
Catalog No. NC2160300
Encompass
Made from high quality stainless steel for optimal thermal conductivity, and they have a well polished surface for easy paraffin block removal. All of the corners are rounded for good paraffin ribboning. These Base Molds are for all applications in specimen embedding with Tissue-Tek Embedding Rings .
This item is not returnable. View return policy
Catalog No. NC2160300 Supplier Electron Microscopy Sciences Supplier No. 6252838
Made from high quality stainless steel for optimal thermal conductivity, and they have a well polished surface for easy paraffin block removal. All of the corners are rounded for good paraffin ribboning. These Base Molds are for all applications in specimen embedding with Tissue-Tek Embedding Rings .