Learn More
Med Vet International Base Mold, 7mm X 7mm X 5mm, 12/Pack
Catalog No. 502831700
Encompass_Preferred
These reusable stainless steel base molds are for use in histology for biopsy. They have extended sides to prevent tipping, provide a great gripping surface for better handling, and to fit with our embedding cassettes for holding paraffin and tissue samples. These molds are available in five different sizes for various uses.
Catalog No. 50-283-1700 Supplier Med Vet International Supplier No. HB07
These reusable stainless steel base molds are for use in histology for biopsy. They have extended sides to prevent tipping, provide a great gripping surface for better handling, and to fit with our embedding cassettes for holding paraffin and tissue samples. These molds are available in five different sizes for various uses.