Learn More
Electron Microscopy Sciences Aluminum Base Mold, 50 x 50 x 30 mm
SDP
Catalog No. 501929772
Encompass
This extra large square base mold offers optimal thermal conductivity. Features a well polished surface for easy block removal. It will accommodate most samples.
Catalog No. 50-192-9772 Supplier Electron Microscopy Sciences Supplier No. 6254715
This extra large square base mold offers optimal thermal conductivity. Features a well polished surface for easy block removal. It will accommodate most samples.