Learn More
Electron Microscopy Sciences Embedding Cassette Metal Base Mold, 37 x 24 mm, 5 mm height
SDP
Catalog No. 501928631
Encompass_Preferred
Fits most standard tissue embedding cassettes. Stainless steel construction provides superior thermal conductivity. The molds have a highly polished surface for easy paraffin block removal and cleaning. The well corners are rounded for effortless tissue block removal and optimal paraffin ribboning.
Footprint: 37 x 24 mm
Height: 5 mm
Catalog No. 50-192-8631 Supplier Electron Microscopy Sciences Supplier No. 7091104
Fits most standard tissue embedding cassettes. Stainless steel construction provides superior thermal conductivity. The molds have a highly polished surface for easy paraffin block removal and cleaning. The well corners are rounded for effortless tissue block removal and optimal paraffin ribboning.
Footprint: 37 x 24 mm
Height: 5 mm