Learn More
Andwin Scientific BASE MOLD RELEASE
SDP
Catalog No. NC9668505
Encompass_Preferred
The base mold release agent is used to coat base molds and detach paraffin from mold. It is packaged in a 4 oz. nonaerosol pump spray bottle and is sold in a case of 12.
This item is not returnable. View return policy
Catalog No. NC9668505 Supplier Andwin Scientific Supplier No. M7316
The base mold release agent is used to coat base molds and detach paraffin from mold. It is packaged in a 4 oz. nonaerosol pump spray bottle and is sold in a case of 12.