Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Base/Neutrals Mix 2 (For EPA 625, CLP)

Catalog No. USUS101N
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-US-101N 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-US-101N Supplier Agilent Technologies Supplier No. US101N1
May include imposed supplier surcharges.
Add to Cart
Add to Cart

† Designates A2LA-Certified product.

Specifications

Chemical Name or Material Base/Neutrals Mix 2
Packaging Ampule
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) Each component at 2000μg/mL in Methylene Chloride:
Azobenzene
2-Chloronaphthalene
1,2-Dichlorobenzene
1,3-Dichlorobenzene
1,4-Dichlorobenzene
2,4-Dinitrotoluene
2,6-Dinitrotoluene
Hexachlorobenzene
Hexachlorobutadiene
Hexachlorocyclopentadiene
Hexachloroethane
Isophorone
Nitrobenzene
1,2,4-Trichlorobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.